Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta7 Armenian Hamster anti-Human, Clone: TN114, MilliporeSigma™

Catalog No. MAB2638MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2638MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2638MI Supplier MilliporeSigma Supplier No. MAB2638
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

Anti-Integrin beta7 Antibody, clone TN114 is an antibody against Integrin beta7 for use in flow cytometry, immunoprecipitation, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin beta7
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone TN114
Conjugate Unconjugated
Formulation Purified armenian hamster monoclonal IgG in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_000880.1
Gene Symbols ITGB7
Host Species Armenian Hamster
Immunogen Purified human integrin beta7 protein.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.