Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin αIIb Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB1967MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1967MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1967MI Supplier MilliporeSigma Supplier No. AB1967
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Integrin αIIb Antibody, cytoplasmic domain is an antibody against Integrin αIIb for use in ELISA and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin αIIb
Applications ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation Whole rabbit serum. Liquid containing no preservative.
Gene Symbols ITGA2B; CD41B; GPIIb; HPA3; GP2B; ITGAB; CD41; GTA
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the cytoplasmic domain of human integrin alphaIIb conjugated to KLH. (a.a. 1020-1039 KVGFFKRNRPPLEEDDEEGE)
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000419.3
Target Species Human
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.