Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin αVΒ3 Mouse anti-Avian, Bovine, Canine, Chicken, Human, Monkey, Pig, Rabbit, AlexaFluor™488, Clone: LM609, MilliporeSigma™

Catalog No. MAB1976XMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1976XMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1976XMI Supplier MilliporeSigma Supplier No. MAB1976X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Integrin αVβ3 Antibody, clone LM609, Alexa Fluor™488 conjugated is validated for use in flow cytometry, immunofluorescence for the detection of Integrin αVβ3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin αVβ3
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone LM609
Conjugate Alexa Fluor 488
Formulation Purified immunoglobulin from Protein A Sepharose™ chromatography, conjugated to Alexa Fluor™ 488. Liquid in 0.02M PB with 0.25M NaCl, pH 7.6, and 0.1% NaN3 as a preservative.
Gene Accession No. NM_002210.2
Gene Symbols ITGAV, DKFZp686A08142, MSK8, CD51, VNRA
Host Species Mouse
Immunogen Purified adhesion receptor from the human melanoma cell line M21 (Cheresh, 1987).
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Avian, Bovine, Canine, Chicken, Human, Monkey, Pig, Rabbit
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.