Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin αVΒ3 Mouse anti-Avian, Bovine, Canine, Chicken, Human, Monkey, Pig, Rabbit, Biotin, Clone: LM609, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1976B
Description
Detect Integrin αVβ3 using this Anti-Integrin αVβ3 Antibody, clone LM609, biotin conjugated validated for use in flow cytometry, immunohistochemistry and immunoprecipitation.
Specifications
Integrin αVβ3 | |
Monoclonal | |
Biotin | |
NM_002210.2 | |
Mouse | |
Protein A purfied | |
Primary | |
2-8°C in undiluted aliquots for up to 6 months. | |
IgG1 |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
LM609 | |
Liquid in 0.01M PBS (pH 7.1), 0.1% sodium azide with 15mg/mL BSA added as a stabilizer. | |
ITGAV, DKFZp686A08142, MSK8, CD51, VNRA | |
Purified adhesion receptor from the human melanoma cell line M21 (Cheresh, 1987). | |
100 μg | |
Avian, Bovine, Canine, Chicken, Human, Monkey, Pig, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction