Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin αVΒ3 Mouse anti-Human, Clone: 27.1 (VNR-1), MilliporeSigma™

Catalog No. MAB1876MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1876MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1876MI Supplier MilliporeSigma Supplier No. MAB1876
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Integrin αVβ3 Antibody, clone 27.1 (VNR-1) detects level of Integrin αVβ3 and has been published and validated for use in IF and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin αVβ3
Applications Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 27.1 (VNR-1)
Conjugate Unconjugated
Formulation Protein A purified mouse immunoglobulin in liquid in 0.02M PB, pH 7.6, 0.25M NaCl containing 0.1% sodium azide.
Gene Accession No. NM_002210.2
Gene Symbols ITGAV, DKFZp686A08142, MSK8, CD51, VNRA
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for up to 12 months in undiluted aliquots.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.