Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin αVΒ3 Mouse anti-Avian, Bovine, Canine, Chicken, Human, Monkey, Pig, Rabbit, Clone: LM609, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1976Z
Description
This Anti-Integrin αVβ3 Antibody, clone LM609, Azide Free is validated for use in flow cytometry, immunohistochemistry, immunoprecipitation, FUNC for the detection of Integrin αVβ3.
Specifications
Integrin αVβ3 | |
Monoclonal | |
Unconjugated | |
NM_002210.2 | |
Mouse | |
Protein A purfied | |
Primary | |
2-8°C in undiluted aliquots for up to 6 months. | |
IgG1 |
Flow Cytometry, Functional Assay, Immunohistochemistry, Immunoprecipitation | |
LM609 | |
Protein A Purified mouse immunoglobulin in 20mM sodium phosphate, 250mM NaCl, pH. 7.6, with no preservatives. | |
ITGAV, DKFZp686A08142, MSK8, CD51, VNRA | |
Purified adhesion receptor from the human melanoma cell line M21 (Cheresh, 1987). | |
100 μg | |
Avian, Bovine, Canine, Chicken, Human, Monkey, Pig, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction