Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin αVΒ3 Mouse anti-Avian, Bovine, Canine, Chicken, Human, Monkey, Pig, Rabbit, Clone: LM609, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1976
Description
Anti-Integrin αVβ3 Antibody, clone LM609 is an antibody against Integrin αVβ3 for use in flow cytometry, immunofluoresence, immunohistochemistry and immunoprecipitation and has published in over 60 citations.
Specifications
Integrin αVβ3 | |
Monoclonal | |
Unconjugated | |
NM_002210.2 | |
Mouse | |
Protein A purfied | |
Primary | |
2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation | |
LM609 | |
Immunoglobulin was Purified by protein A affinity and is presented as a liquid in 0.02M PB, pH 7.6, 0.25M NaCl containing 0.1% sodium azide. | |
ITGAV, DKFZp686A08142, MSK8, CD51, VNRA | |
Purified adhesion receptor from the human melanoma cell line M21 (Cheresh, 1987). | |
100 μg | |
Avian, Bovine, Canine, Chicken, Human, Monkey, Pig, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction