Learn More
MilliporeSigma™ anti-Integrin αVβ3 (Vitronectin Receptor; CD51/CD61), Clone: LM609, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB1976/DEL
Description
Cheresh, 1987.
Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Specifications
Integrin αVβ3 | |
Unconjugated | |
Purified adhesion receptor from the human melanoma cell line M21 | |
RUO | |
Primary | |
Semi-purified |
Monoclonal | |
Mouse | |
100 μg | |
Cell Structure | |
Avian, Bovine, Canine, Chicken, Human, Monkey, Pig, Rabbit | |
IgG1 |