Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin αVΒ5 Mouse anti-Human, Clone: 15F11, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB2019Z
Description
Anti-Integrin αVβ5 Antibody, clone 15F11, Azide Free is an antibody against Integrin αVβ5 for use in ELISA, flow cytometry, immunoprecipitation and immunocytochemistry.
Specifications
Integrin αVβ5 | |
Monoclonal | |
Unconjugated | |
NM_002210.2 | |
Mouse | |
100 μg | |
Human | |
Purified |
ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation | |
15F11 | |
Purified immunoglobulin. Liquid at 1mg/mL in PBS, containing no preservatives. | |
ITGAV, DKFZp686A08142, MSK8, CD51, VNRA | |
Generated against human integrin alphaVbeta5 purified from placental tissue and depleted of integrin alphaVbeta3. | |
Primary | |
2-8°C in undiluted aliquots for up to 12 months. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction