Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin αV Mouse anti-Human, Clone: AV1, MilliporeSigma™

Catalog No. MAB2021ZMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2021ZMI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2021ZMI Supplier MilliporeSigma Supplier No. MAB2021Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Integrin αV Antibody, extracellular, clone AV1, azide free detects level of Integrin αV and has been published and validated for use in FC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin αV
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone AV1
Conjugate Unconjugated
Formulation Unpurified tissue culture supernatant from a perfusion system, filtered through a 0.2μ micron membrane prior to vialing. Product contains 20%FBS and Ciprofloxacin at final concentration of 10μg/mL.
Gene Accession No. NM_002210.2
Gene Symbols ITGAV, DKFZp686A08142, MSK8, CD51, VNRA
Host Species Mouse
Immunogen Human integrin alphaVbeta3 heterodimer isolated from placenta.
Quantity 500 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.