Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin αV Mouse anti-Human, Phycoerythrin, Clone: 13C2, MilliporeSigma™

Catalog No. CBL490PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL490PMI 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL490PMI Supplier MilliporeSigma Supplier No. CBL490P

Mouse Monoclonal Antibody

Detect Integrin αV using this Anti-Integrin αV Antibody, clone 13C2, Phycoerythrin conjugated validated for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin αV
Applications Flow Cytometry
Classification Monoclonal
Clone 13C2
Conjugate PE
Formulation The conjugate is supplied as a 100 test vial in phosphate buffered saline containing 10mM sodium azide and 1mg/ml bovine serum albumin. For flow cytometry we recommend using 10μL of conjugate per test.
Gene Accession No. NM_002210.2
Gene Symbols ITGAV, DKFZp686A08142, MSK8, CD51, VNRA
Host Species Mouse
Immunogen Human osteoclastoma tissue
Quantity 1 Unit
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C, protect from prolonged exposure to light. Do not Freeze.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.