Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interferon-Β Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB1431MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20,000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1431MI 20,000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1431MI Supplier MilliporeSigma Supplier No. AB1431
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Interferon-β Antibody is validated for use in ELISA, immunohistochemistry, immunoprecipitation, NEUT, western blotting for the detection of Interferon-β.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Interferon-β
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Neutralization, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit Serum diluited with PBS. Liquid containing no preservatives.
Gene Symbols IL6; HSF; BSF-2; HGF; IL-6; BSF2; CDF; IFNB2
Host Species Rabbit
Immunogen Recombinant human interferon beta expressed in E. coli
Purification Method Unpurified
Quantity 20,000 U
Primary or Secondary Primary
Gene ID (Entrez) NM_000600.1
Target Species Human
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.