Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interferon-Β Mouse anti-Human, Clone: A1, MilliporeSigma™

Catalog No. MAB416MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB416MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB416MI Supplier MilliporeSigma Supplier No. MAB416
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Interferon-β Antibody, clone A1 detects level of Interferon-β and has been published and validated for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Interferon-β
Applications ELISA, Western Blot
Classification Monoclonal
Clone A1
Conjugate Unconjugated
Formulation Immunoglobulin purified by Protein A from culture supernatant. Liquid in Phosphate Buffered Saline, pH 7.2
Gene Accession No. NM_000600.2
Gene Symbols IL6, IL-6, HSF, BSF2, BSF-2, CDF, IFNB2, HGF
Host Species Mouse
Immunogen Human interferon beta.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.