Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interferon-γ Rat anti-Mouse, Clone: RMMG-1, MilliporeSigma™

Catalog No. MAB1152MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1152MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1152MI Supplier MilliporeSigma Supplier No. MAB1152
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Interferon-γ Antibody, clone RMMG-1 is an antibody against Interferon-γ for use in ELISA, immunoprecipitation, western blot, immunohistochemistry, NEUT.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Interferon-γ
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Neutralization, Western Blot
Classification Monoclonal
Clone RMMG-1
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS containing 0.1% bovine serum albumin.
Gene Symbols IFNG
Host Species Rat
Immunogen Mouse Interferon gamma
Quantity 500 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.