Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interferon Lambda 4 Mouse anti-Human, Non-human Primate, Clone: 4G1, MilliporeSigma™

Catalog No. MABF227MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF227MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF227MI Supplier MilliporeSigma Supplier No. MABF227
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Interferon Lambda 4 antibody, clone 4G1 is validated for use in ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, and Western Blotting for the detection of Interferon Lambda 4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Interferon Lambda 4
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4G1
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001263183
Gene Symbols IFNL4
Host Species Mouse
Immunogen Linear peptide corresponding to human Interferon Lambda 4.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.