Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interferon-α/Β Receptor Chain 2 Mouse anti-Human, Clone: MMHAR-2, MilliporeSigma™

Catalog No. MAB1155MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1155MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1155MI Supplier MilliporeSigma Supplier No. MAB1155
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Interferon-α/β Receptor Chain 2 Antibody, clone MMHAR-2 is an antibody against Interferon-α/β Receptor Chain 2 for use in flow cytometry, immunoprecipitation, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Interferon-α/β Receptor Chain 2
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone MMHAR-2
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS containing 0.1% bovine serum albumin (BSA).
Gene Accession No. NM_000605.2
Gene Symbols IFNA2, INFA2, IFNA, MGC125764, MGC125765
Host Species Mouse
Immunogen Human Interferon alpha/beta Receptor Chain 2
Quantity 50 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.