Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interleukin-2 (IL-2) Mouse anti-Human, Clone: AB12.38.35C3.3F9, MilliporeSigma™

Catalog No. 041584MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-158-4MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-158-4MI Supplier MilliporeSigma Supplier No. 041584
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Interleukin-2 (IL-2) using this Anti-Interleukin-2 (IL-2) Antibody, clone 3F9 validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Interleukin-2 (IL-2)
Applications Western Blot
Classification Monoclonal
Clone AB12.38.35C3.3F9
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_000577
Gene Symbols IL2, IL-2, TCGF
Host Species Mouse
Immunogen Full length recombinant protein corresponding to human IL-2.
Purification Method Protein G Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.