Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interleukin 21 (IL-21) Mouse anti-Human, Clone: AB43.50.32H5.1G8, MilliporeSigma™

Catalog No. 041583MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-158-3MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-158-3MI Supplier MilliporeSigma Supplier No. 041583
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Interleukin 21 (IL-21) Antibody, clone 1G8 is validated for use in western blot, ELISA for the detection of Interleukin 21 (IL-21).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Interleukin 21 (IL-21)
Applications ELISA, Western Blot
Classification Monoclonal
Clone AB43.50.32H5.1G8
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_068575
Gene Symbols Za11, IL-21, IL21
Host Species Mouse
Immunogen Full length recombinant protein corresponding to human IL-21.
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.