Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interleukin 23 (IL-23) p40 Mouse anti-Human, Clone: AB54.69.3G2.1D9, MilliporeSigma™

Catalog No. 041582MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-158-2MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-158-2MI Supplier MilliporeSigma Supplier No. 041582

Mouse Monoclonal Antibody

Anti-Interleukin 23 (IL-23) p40 Antibody, clone 2G6 detects level of Interleukin 23 (IL-23) p40 and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Interleukin 23 (IL-23) p40
Applications Western Blot
Classification Monoclonal
Clone AB54.69.3G2.1D9
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_002178
Gene Symbols IL-23-A, SGRF, IL-23p19, IL23P19, P19, IL-23A, IL-23
Host Species Mouse
Immunogen Full length cohesin-tagged recombinant protein corresponding to human IL-23 protein.
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.