Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Interleukin-6 Rabbit anti-Sheep, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB1839
Description
Anti-Interleukin-6 Antibody is an antibody against Interleukin-6 for use in ELISA and western blotting.
Specifications
Interleukin-6 | |
Polyclonal | |
Rabbit serum. Liquid containing 0.1% sodium azide. | |
Rabbit | |
100 μL | |
NM_000600.2 | |
−20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles. |
ELISA, Western Blot | |
Unconjugated | |
IL6; HSF; BSF-2; HGF; IL-6; BSF2; CDF; IFNB2 | |
Recombinant ovine IL-6 | |
Primary | |
Sheep | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction