Learn More
Description
Specifications
Specifications
| Antigen | Interleukin-6 |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry, Neutralization, Western Blot |
| Classification | Monoclonal |
| Clone | B-E8 |
| Conjugate | Unconjugated |
| Formulation | The monoclonal is purified from tissue culture supernatant by ion exchange chromatography and is presented as a liquid in phosphate buffered saline, pH 7.2, containing 0.09% sodium azide. |
| Gene Accession No. | NM_000600.2 |
| Gene Symbols | IL6, HSF, BSF-2, HGF, IL-6, BSF2, CDF, IFNB2 |
| Host Species | Mouse |
| Immunogen | Recombinant human IL-6. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.