Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interleukin-6 (IL-6) Mouse anti-Human, Clone: AB42_51.38C8.1G3, MilliporeSigma™

Catalog No. MABF41MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF41MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF41MI Supplier MilliporeSigma Supplier No. MABF41
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Interleukin-6 (IL-6) Antibody, clone 1G3 is an antibody against Interleukin-6 (IL-6) for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Interleukin-6 (IL-6)
Applications Western Blot
Classification Monoclonal
Clone AB42_51.38C8.1G3
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_000591
Gene Symbols IL6, IL-6, CDF, HSF, HGF, BSF2, IFNB2, BSF-2
Host Species Mouse
Immunogen Human IL-6 fused to the soluble domain of the IL-6R.
Purification Method Protein G Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.