Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interleukin-8 Rabbit anti-Sheep, Polyclonal, MilliporeSigma™

Catalog No. AB1840MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1840MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1840MI Supplier MilliporeSigma Supplier No. AB1840
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Interleukin-8 Antibody detects level of Interleukin-8 and has been published and validated for use in ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Interleukin-8
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit serum. Liquid containing 0.1% sodium azide.
Gene Symbols IL8; GCP-1; emoctakin; MDNCF; NAP1; CXCL8; AMCF-I; IL-8; Emoctakin; TSG-1; NAP-1; LYNAP; MONAP; SCYB8; GCP1; LUCT; LECT; K60; 3-10C; b-ENAP; NAF
Host Species Rabbit
Immunogen Recombinant ovine IL-8
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000584.2
Target Species Sheep
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.