Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interleukin-8 Mouse anti-Bovine, Goat, Sheep, Clone: 8M6, MilliporeSigma™

Catalog No. MAB1044MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1044MI 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1044MI Supplier MilliporeSigma Supplier No. MAB1044
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Interleukin-8 Antibody, clone 8M6 is validated for use in ELISA, flow cytometry, NEUT, western blotting for the detection of Interleukin-8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Interleukin-8
Applications ELISA, Flow Cytometry, Neutralization, Western Blot
Classification Monoclonal
Clone 8M6
Conjugate Unconjugated
Formulation Liquid in PBS containing 0.1% sodium azide.
Gene Accession No. NM_000584.2
Gene Symbols IL8, GCP-1, emoctakin, MDNCF, NAP1, CXCL8, AMCF-I, IL-8, Emoctakin, TSG-1, NAP-1, LYNAP, MONAP, SCYB8, GCP1, LUCT, LECT, K60, 3-10C, b-ENAP, NAF
Host Species Mouse
Immunogen Recombinant ovine IL-8
Quantity 250 μg
Primary or Secondary Primary
Target Species Bovine, Goat, Sheep
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.