Learn More
Internexin α Rabbit anti-Mammals, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5354
Description
Anti-Internexin Antibody, α is an antibody against Internexin for use in ELISA, western blot, immunocytochemistry, immunohistochemistry.
Specifications
Internexin α | |
Polyclonal | |
Serum liquid, containing no preservatives. | |
Rabbit | |
50 μL | |
NM_032727.2 | |
−20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
Unconjugated | |
INA; NF-66; NEF5; MGC12702; Alpha-Inx; TXBP-1; Neurofilament-66; Alpha-internexin | |
Full length recombinant rat alpha-internexin fused to E. coli Trp E. | |
Primary | |
Mammalia | |
Serum |