Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IP-10 Mouse anti-Human, Unlabeled, Clone: 4D5, BD

Catalog No. BDB555046 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555046 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555046 Supplier BD Biosciences Supplier No. 555046
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The monoclonal antibody 4D5/A7/C5 reacts with human CXC chemokine, interferon gamma inducible protein (IP-10). IP-10 is inducible in monocytes, keratinocytes, and endothelial cells by IFN-γ. This antibody is routinely tested by ELISA Capture. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen IP-10
Applications ELISA
Classification Monoclonal
Clone 4D5
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human IP-10 protein
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.