Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IP-10 Mouse anti-Human, PE, Clone: 6D4/D6/G2, BD

Catalog No. BDB555049 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555049 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555049 Supplier BD Biosciences Supplier No. 555049
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The monoclonal antibody 6D4/D6/G2 reacts with human CXC chemokine, interferon gamma inducible protein 10 (IP-10). IP-10 is inducible in monocytes, keratinocytes and endothelial cells by IFN-γ. The immunogen used to generate the monoclonal antibody 6D4/D6/G2 was recombinant human IP-10 protein. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen IP-10
Applications Flow Cytometry
Classification Monoclonal
Clone 6D4/D6/G2
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human IP-10 protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.