Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IP3 Receptor 1 Rabbit anti-Mouse, Human, Canine, Rat, Polyclonal, MilliporeSigma™

Catalog No. 071213MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-121-3MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-121-3MI Supplier MilliporeSigma Supplier No. 071213
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-IP3 Receptor 1 Antibody is validated for use in immunohistochemistry (paraffin), western blotting for the detection of IP3 Receptor 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IP3 Receptor 1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit serum. Liquid
Gene Symbols INSP3R1; IP3R; IP3R1; InsP3R1; Insp3r1; SCA15; SCA16
Host Species Rabbit
Immunogen Synthetic peptide from the N-terminal of human IP3 Receptor 1.
Purification Method Unpurified rabbit serum
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) Q14643
Target Species Canine, Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Isotype IgG Mix
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.