Learn More
Description
Specifications
Specifications
| Antigen | IP3 Receptor 1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, 5% sucrose as a stabilizer and 0.025% sodium azide as a preservative. Reconstitute with 200μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg fo |
| Gene Symbols | ITPR1; IP3R1; SCA15; InsP3R1; IP3R; INSP3R1; Insp3r1 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding amino acids 2732-2750 of rat IP3R1 (Accession number P29994). It is expected that the antibody will also react with mouse and human due to identical sequence homology. |
| Purification Method | Affinity Purified |
| Quantity | 200 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.