Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IP3 Receptor 2 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB9074MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9074MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9074MI Supplier MilliporeSigma Supplier No. AB9074
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect IP3 Receptor 2 using this Anti-IP3 Receptor 2 Antibody validated for use in immunocytochemistry, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IP3 Receptor 2
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit serum. Liquid.
Gene Symbols ITPR2; InsP3R2; IP3R2
Host Species Rabbit
Immunogen Synthetic peptide from the C terminus of human IP3 Receptor 2.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002223.2
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.