Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IP3 Receptor Rabbit anti-Bovine, Human, Mouse, Pig, Rabbit, Rat, Canine, Polyclonal, MilliporeSigma™

Catalog No. 071210MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-121-0MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-121-0MI Supplier MilliporeSigma Supplier No. 071210
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-IP3 Receptor Antibody is validated for use in western blot, immunofluoresence, immunohistochemistry (paraffin), ELISA, immunoprecipitation, RIA for the detection of IP3 Receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IP3 Receptor
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in buffer containing 10mM sodium phosphate, 150mM NaCl, and 0.01% sodium azide, pH 7.5.
Gene Symbols ITPR1; IP3R1; Insp3r1; IP3R; INSP3R1; SCA15; InsP3R1
Host Species Rabbit
Immunogen Synthetic peptide (KDSTEYTGPESYV) corresponding to the C-terminus of human IP3 Receptor type 1.
Purification Method Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001099952.1
Target Species Bovine, Canine, Human, Mouse, Pig, Rabbit, Rat
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.