Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IP3R3 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1002545
Description
IP3R3 Polyclonal specifically detects IP3R3 in Human, Mouse, Rat, Bovine samples. It is validated for Peptide ELISA.Specifications
| IP3R3 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| FLJ36205, inositol 14,5-triphosphate receptor, type 3, inositol 14,5-trisphosphate receptor type 3, InsP3R3, IP3 receptor, IP3 receptor isoform 3, IP3R, IP3R 3, IP3R3, Type 3 inositol 14,5-trisphosphate receptor, Type 3 InsP3 receptor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3710 | |
| Human, Mouse, Rat, Bovine | |
| IgG |
| ELISA | |
| 0.5 mg/mL | |
| Peptide ELISA Detection limit 1:128000 | |
| NP_002215.1 | |
| ITPR3 | |
| Peptide with sequence C-DKKERPTDEEGFLH corresponding to internal region according to NP_002215.2. | |
| 0.1 mg | |
| Primary | |
| No cross-reactivity expected with other types of ITPR | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction