Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IQGAP1 Mouse anti-Human, Clone: AF4, MilliporeSigma™

Catalog No. 05504MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-504-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-504-MI Supplier MilliporeSigma Supplier No. 05504
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect IQGAP1 using this Anti-IQGAP1 Antibody, clone AF4 validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IQGAP1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone AF4
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Gene Accession No. NM_003870
Gene Symbols IQGAP1, HUMORFA01, SAR1, KIAA0051, p195
Host Species Mouse
Immunogen A Glu-Glu tagged C-terminal fragment corresponding to amino acids 863-1657 of human IQGAP1, purified from baculovirus infected insect cells
Purification Method Protein G chromatography
Quantity 200 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.