Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRAK1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 071601MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-160-1MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-160-1MI Supplier MilliporeSigma Supplier No. 071601
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-IRAK1 Antibody, C-terminus is validated for use in western blotting for the detection of IRAK1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRAK1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS and 0.09% sodium azide.
Gene Symbols IRAK; IRAK-1 ; pelle; EC 2.7.11.1
Host Species Rabbit
Immunogen KLH-conjugated synthetic linear peptide corresponding to the C-terminal region of IRAK1.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_001020413.1
Target Species Human
Content And Storage 2-8°C in undiluted aliquotes for up to 6 months, −20°C for long term. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.