Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

interleukin-1 receptor-associated kinase 1 binding protein 1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016616 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-616 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-616 Supplier Abnova Corporation Supplier No. H00134728B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human IRAK1BP1 protein.

Sequence: MSLQKTPPTRVFVELVPWADRSRENNLASGRETLPGLRHPLSSTQAQTATREVQVSGTSEVSAGPDRAQVVVRVSSTKEAAAEAKKSVCRRLDYITQSLQQQGVQAENITVTKDFRRVENAYHMEAEVCITFTEFGKMQNICNFLVEKLDSSVVISPPQFYHTPGSVENLRRQACLVAVENAWRKAQEVCNLVGQTLGKPLLIKEEETKEWEGQIDDHQSSRLSSSLTVQQKIKSATIHAASKVFITFEVKGKEKRKKHL

Specifications

Antigen interleukin-1 receptor-associated kinase 1 binding protein 1
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human IRAK1BP1 protein.
Formulation PBS with no preservative; pH 7.4
Gene IRAK1BP1
Gene Accession No. NM_001010844.1
Gene Alias AIP70/MGC138458/MGC138460/SIMPL
Gene Symbols IRAK1BP1
Host Species Mouse
Immunogen IRAK1BP1 (NP_001010844.1, 1 a.a. ∼ 260 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Kinases
Primary or Secondary Primary
Gene ID (Entrez) 134728
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.