Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
interleukin-1 receptor-associated kinase 3, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human IRAK3 protein.
Supplier: Abnova Corporation H00011213D01P
Description
Sequence: MAGNCGARGALSAHTLLFDLPPALLGELCAVLDSCDGALGWRGLAERLSSSWLDVRHIEKYVDQGKSGTRELLWSWAQKNKTIGDLLQVLQEMGHRRAIHLITNYGAVLSPSEKSYQEGGFPNILFKETANVTVDNVLIPEHNEKGVLLKSSISFQNIIEGTRNFHKDFLIGEGEIFEVYRVEIQNLTYAVKLFKQEKKMQCKKHWKRFLSELEVLLLFHHPNILELAAYFTETEKFCLIYPYMRNGTLFDRLQCVGDTAPLPWHIRIGILIGISKAIHYLHNVQPCSVICGSISSANILLDDQFQPKLTDFAMAHFRSHLEHQSCTINMTSSSSKHLWYMPEEYIRQGKLSIKTDVYSFGIVIMEVLTGCRVVLDDPKHIQLRDLLRELMEKRGLDSCLSFLDKKVPPCPRNFSAKLFCLAGRCAATRAKLRPSMDEVLNTLESTQASLYFAEDPPTSLKSFRCPSPLFLENVPSIPVEDDESQNNNLLPSDEGLRIDRMTQKTPFECSQSEVMFLSLDKKPESKRNEEACNMPSSSCEESWFPKYIVPSQDLRPYKVNIDPSSEAPGHSCRSRPVESSCSSKFSWDEYEQYKKESpecifications
interleukin-1 receptor-associated kinase 3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human IRAK3 protein. | |
IRAK3 | |
ASRT5/FLJ13601/IRAK-M/IRAKM | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_007199.1 | |
IRAK3 | |
IRAK3 (NP_009130.1, 1 a.a. ∼ 596 a.a) full-length human protein. | |
100 μg | |
Kinases | |
11213 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction