Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRAK4 Mouse anti-Human, PE, Clone: L29-525, BD

Catalog No. BDB560303 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560303 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560303 Supplier BD Biosciences Supplier No. 560303
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Interleukin 1 (IL-1) is a proinflammatory cytokine that mediates the host inflammatory response. IL-1 exerts its biological effects through its type I receptor (IL-1RI), which is found on most cell types. When IL-1 binds to the IL-1RI, NF-κB (transcription factor) is activated and translocates to the nucleus where it induces gene expression. IL-1-receptor-associated kinase (IRAK) has been reported to co-immunoprecipitate with IL-1RI. The IRAK family is made up of four identified Pelle homologs that regulate the activation of NF-κB and mitogen-activated protein (MAP) kinase pathways. IRAK4 is a unique member of the IRAK family because it is the closest mammalian homolog to Pelle. Also, when overexpressed, its kinase activity allows it to activate both the NF-κB and mitogen-activated protein (MAP) kinase pathways, and block the IL-1-induced activation and modification of IRAK1. IRAK4 is also able to phosphorylate IRAK1, which suggests that IRAK4 affects the early signal transduction of Toll/IL-1 receptors before the involvement of IRAK1. The L29-525 monoclonal antibody recognizes human IRAK4, regardless of phosphorylation status. The specificity of this antibody conjugate for flow cytometric analysis was validated by confirming that RNA-mediated interference (RNAi) of the specific protein reduced the staining of the cells (see figure). Furthermore, the capacity of the RNAi to down-regulate the expression of the relevant protein was confirmed by western blot analysis.

Host Species: Mouse
Clone: L29-525
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human IRAK4 Recombinant Protein

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen IRAK4
Applications Flow Cytometry
Classification Monoclonal
Clone L29-525
Conjugate PE
Description IRAK-4, REN64, Q69FE3
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human IRAK4 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.