Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF-7 Mouse anti-Human, Alexa Fluor 488, Clone: K40-321, BD

Catalog No. BDB558707 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558707 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558707 Supplier BD Biosciences Supplier No. 558707
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Interferon regulatory factor 7 (IRF-7) is a transcription factor that regulates anti-viral defenses by controlling the induction of type-I interferon (IFN) responses. IRF-7 expression is induced in lymphoid cells by virus infection, as well as by IFN, lipopolysaccharide, and TNF-α. IRF-7 responses are initiated by Toll-like receptors (TLR) or the cytoplasmic protein retinoic acid inducible gene I (RIG-I). Upon TLR activation, it forms cytoplasmic complexes with MyD88, an adaptor in the TLR signaling pathways. The TLR-dependent and RIG-I-dependent pathways activate kinases, such as IKK-ε and TBK1, that phosphorylate IRF-7 and induce movement of IRF-7-containing complexes to the nucleus, where it preferentially activates IFN-α promoters. The K40-321 monoclonal antibody recognizes human IRF-7, regardless of phosphorylation status. Our in-house testing is performed on a cell line that has been co-transfected with TBK1 and IRF-7. Phosphorylation of IRF-7 in the transfectants requires virus infection or over-expression of a signaling molecule of the RIG-I pathway, such as TBK1. Phosphorylation of endogenous IRF-7 in untransfected cells has not yet been detected. We confirmed that mAb K40-321 does not cross-react with TBK1 by Western blot analysis using the purified antibody.

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF-7
Applications Flow Cytometry
Classification Monoclonal
Clone K40-321
Conjugate Alexa Fluor 488
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human IRF-7 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.