Learn More
IRF-7 Mouse anti-Human, PE, Clone: K40-321, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 558706
Description
Interferon regulatory factor 7 (IRF-7) is a transcription factor that regulates anti-viral defenses by controlling the induction of type-I interferon (IFN) responses. IRF-7 expression is induced in lymphoid cells by virus infection, as well as by IFN, lipopolysaccharide, and TNF-α. IRF-7 responses are initiated by Toll-like receptors (TLR) or the cytoplasmic protein retinoic acid inducible gene I (RIG-I). Upon TLR activation, it forms cytoplasmic complexes with MyD88, an adaptor in the TLR signaling pathways. The TLR-dependent and RIG-I-dependent pathways activate kinases, such as IKK-ε and TBK1, that phosphorylate IRF-7 and induce movement of IRF-7-containing complexes to the nucleus, where it preferentially activates IFN-α promoters. The K40-321 monoclonal antibody recognizes human IRF-7, regardless of phosphorylation status. Our in-house testing is performed on a cell line that has been co-transfected with TBK1 and IRF-7. Phosphorylation of IRF-7 in the transfectants requires virus infection or over-expression of a signaling molecule of the RIG-I pathway, such as TBK1. Phosphorylation of endogenous IRF-7 in untransfected cells has not yet been detected. We confirmed that mAb K40-321 does not cross-react with TBK1 by Western blot analysis using the purified antibody.
Intracellular Staining

Specifications
IRF-7 | |
Monoclonal | |
PE | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
K40-321 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human IRF-7 Recombinant Protein | |
50 Tests | |
Cell Biology | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.