Learn More
IRGM Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier:  MilliporeSigma ABC338
Catalog No. ABC338MI
Only null left
Add to Cart
Description
Description
Detect Immunity-related GTPase family M protein using this rabbit polyclonal antibody, Anti-IRGM Antibody validated for use in western blotting, immunohistochemistry (Paraffin) and Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IRGM
Polyclonal
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.
Rabbit
Affinity Purified
Primary
2-8°C for 12 months
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
IRGM
KLH-conjugated linear peptide corresponding to the intermediate domain of human IRGM.
100 μg
Human
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only