Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRGQ Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524856
Description

Specifications
IRGQ | |
Polyclonal | |
IRGQ | |
IRGQ1, immunity-related GTPase family Q protein, immunity-related GTPase family, Q1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
126298 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q8WZA9 | |
IRGQ | |
KLH conjugated synthetic peptide between 14-43 amino acids from the N-terminal region of human IRGQ | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction