Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron Pegulatory Protein 1 Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB15506MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB15506MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB15506MI Supplier MilliporeSigma Supplier No. AB15506
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Iron Pegulatory Protein 1 Antibody detects level of Iron Pegulatory Protein 1 and has been published and validated for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Iron Pegulatory Protein 1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid in PBS with 0.1% BSA.
Gene Symbols ACO1; Aconitase; IREBP; IRP1; OTTHUMP00000045233; IREB1; EC 4.2.1.3
Host Species Rabbit
Immunogen Synthetic peptide near the N-terminus of rat IRP-1.
Purification Method Immunoaffinity purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_002197.1
Target Species Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.