Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron Regulatory Protein 2 Mouse anti-Human, MilliporeSigma™

Catalog No. MAB5582MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5582MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5582MI Supplier MilliporeSigma Supplier No. MAB5582

Mouse Monoclonal Antibody

Anti-Iron Regulatory Protein 2 Antibody detects level of Iron Regulatory Protein 2 and has been published and validated for use in ELISA, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Iron Regulatory Protein 2
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Mouse ascites. Liquid. Contains no preservative.
Gene Accession No. NM_004136.2
Gene Symbols IREB2, IRP2AD, ACO3, FLJ23381, IRP2
Host Species Mouse
Immunogen Recombinant fusion protein.
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.