Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron Regulatory Protein 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB15508MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB15508MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB15508MI Supplier MilliporeSigma Supplier No. AB15508
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Iron Regulatory Protein 2 using this Anti-Iron Regulatory Protein 2 Antibody validated for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Iron Regulatory Protein 2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid in PBS with 0.1% BSA and 0.05% sodium azide.
Gene Symbols IREB2; IRP2AD; FLJ23381; IRP2; ACO3
Host Species Rabbit
Immunogen Synthetic peptide near the N-terminus of human/rat/mouse IRP-2.
Purification Method Immunoaffinity purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_004136.2
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.