Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRS-1 Mouse, Unlabeled, Clone: 6, BD

Catalog No. BDB611394 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611394 50 μg
BDB611395 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611394 Supplier BD Biosciences Supplier No. 611394
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The IRS (Insulin receptor substrate) proteins IRS-1, IRS-2, IRS-3, and IRS-4 are major substrates of the insulin receptor tyrosine kinase and the insulin-like growth factor-1 receptor. IRS proteins contain an N-terminal pleckstrin homology (PH) domain, an ATP-binding domain, and multiple tyrosine phosphorylation sites in the C-terminus. Following insulin receptor ligation, IRS-1 binds to the juxtamembrane region of the receptor and is tyrosine phosphorylated. This facilitates its interaction with SH2 domain-containing signaling proteins, such as PI3 kinase, fyn, Grb2, and PTP1D. Phosphorylation dramatically reduces the affinity of IRS-1 for the insulin receptor, indicating that dissociation from the receptor and subsequent subcellular translocation are important to IRS-1 function in the pleiotropic effects induced by insulin. In support of this, IRS-1-null mice are viable, but exhibit growth retardation and abnormal glucose metabolism. In cases of reduced IRS-1 expression, certain IRS-1 functions can be assumed by the related IRS-2 protein, while other activities linked to IRS-1 are inhibited. Thus, IRS-1 is an essential component of insulin induced signal transduction.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen IRS-1
Applications Western Blot
Classification Monoclonal
Clone 6
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat IRS-1 aa. 1131-1234
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.