Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRS-1 (pY896) Mouse, PE, Clone: K9-211, BD

Catalog No. BDB558440 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558440 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558440 Supplier BD Biosciences Supplier No. 558440
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The IRS (Insulin Receptor Substrate) proteins IRS-1, IRS-2, IRS-3, and IRS-4 are major substrates of the insulin receptor and the insulin-like growth factor-1 (IGF-1) receptor tyrosine kinases. IRS proteins contain an N-terminal pleckstrin homology (PH) domain, a phosphotyrosine-binding (PTB) domain, and multiple tyrosine phosphorylation sites in the C-terminus. The IRS-1 protein is widely expressed and, along with IRS-2, mediates somatic growth and carbohydrate metabolic responses to insulin. Following insulin receptor ligation, IRS-1 binds to the juxtamembrane region of the receptor via the PH and PTB domains and is tyrosine phosphorylated, which facilitates its interaction with SH2 domain-containing signaling proteins. Specifically, the phosphorylated tyrosine 896 (pY896) of human IRS-1 is a major binding site for the GRB2 (Growth-factor Receptor-Bound protein 2) adaptor protein. After IRS-1 activation, negative and positive feedback regulates dephosphorylation of its tyrosine sites, which ultimately regulates the magnitude and/or duration of the downstream pleiotropic responses to insulin and IGF-1. The K9-211 monoclonal antibody recognizes pY896 of human IRS-1. The orthologous phosphorylation sites of mouse and rat IRS-1 are Y891 and Y895, respectively.

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen IRS-1 (pY896)
Applications Flow Cytometry
Classification Monoclonal
Clone K9-211
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human IRS-1 Peptide
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.