Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRS-2 Mouse anti-Mouse, Rat, Clone: 9.5.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS15
Description
Anti-IRS-2 Antibody, clone 9.5.2 detects level of IRS-2 and has been published and validated for use in western blotting.
Specifications
IRS-2 | |
Monoclonal | |
Unconjugated | |
NP_001074681.1 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Western Blot | |
9.5.2 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
IRS2, IRS-2 | |
KLH-conjugated linear peptide corresponding to mouse IRS-2. | |
100 μg | |
Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction