Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 06248MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-248-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-248-MI Supplier MilliporeSigma Supplier No. 06248
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-IRS1 Antibody is an antibody against IRS1 for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRS1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified Rabbit IgG in buffer containing 0.2M Trisglycine, pH 7.4, 0.15M NaCl, 0.1mM EDTA, 5mg/mL BSA, and 0.05% sodium azide.
Gene Symbols IRS1; IRS-1; HIRS-1
Host Species Rabbit
Immunogen Carboxy-terminal 14 amino acid peptide ([C]YASINFQKQPEDRQ) of rat liver IRS1
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_005535
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.