Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRS1 Mouse anti-Human, Mouse, Rat, Monkey, Bovine, Canine, Pig, Clone: 4.2.2, MilliporeSigma™

Catalog No. 051085MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-108-5MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-108-5MI Supplier MilliporeSigma Supplier No. 051085
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-IRS1 Antibody, clone 4.2.2 is validated for use in western blot, immunoprecipitation, IC for the detection of IRS1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRS1
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4.2.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody in buffer containing 0.1MTris-Glycine (pH 7.4),150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_005535.1
Gene Symbols HIRS-1 , IRS-1
Host Species Mouse
Immunogen Synthetic peptide corresponding to amino acids 431-446 of mouse IRS1.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Canine, Human, Monkey, Mouse, Pig, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.