Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRS1 Rabbit anti-Mouse, Rat, Human, Clone: 58-10C-31, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma 05784R
Description
Detect IRS1 using this Anti-IRS1 Antibody, clone 58-10C-31 validated for use in western blotting and immunoprecipitation.
Specifications
IRS1 | |
Monoclonal | |
Unconjugated | |
NP_0371 | |
KLH- conjugated - linear peptide corresponding to the C-terminus of rat IRS1. | |
100 μL | |
IRS1, IRS-1, HIRS-1 | |
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles. | |
IgG |
Immunoprecipitation, Western Blot | |
58-10C-31 | |
Purified Rabbit Monoclonal IgG Supernatant in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide and 40% glycerol. | |
Rabbit | |
Protein A purfied | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction